APO455834693 Apollo Scientific Ltd

4-Bromo-2-fluorocinnamic acid

  • MDL number MFCD00143267
  • Cas Number : 149947-19-3
  • Molecular Weight : 245.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 216-221
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-2-fluorocinnamic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-2-fluorocinnamic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart