APO455834696 Apollo Scientific Ltd

Ethyl 2-chloro-4-(trifluoromethyl)-1,3-thiazole-5-carboxylate

  • MDL number MFCD02956778
  • Cas Number : 72850-52-3
  • Molecular Weight : 259.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 57-59
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-2-chloro-4-trifluoromethyl-13-thiazole-5-carboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-2-chloro-4-trifluoromethyl-13-thiazole-5-carboxylate
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart