APO455834698 Apollo Scientific Ltd

4-Bromo-2-fluoro-6-nitrophenol

  • MDL number MFCD00013372
  • Cas Number : 320-76-3
  • Molecular Weight : 236

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63-65
Synonym 5-Bromo-3-fluoro-2-hydroxynitrobenzene
Purity 97
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-2-fluoro-6-nitrophenol

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-2-fluoro-6-nitrophenol
pictograms GHS07
hstatements H302+H312+H332;H319
Availability details for
Quantity
  • Availability
  • Add to Cart