APO455834707 Apollo Scientific Ltd

2-Fluoro-N-methylaniline

  • MDL number MFCD00028030
  • Cas Number : 1978-38-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (2-Fluorophenyl)methylamine
Density 1.1
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9796

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=9796
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart