APO455834711 Apollo Scientific Ltd

3-Amino-5-(trifluoromethyl)-1H-indazole

  • MDL number MFCD01719740
  • Cas Number : 2250-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114-118
Synonym 5-(Trifluoromethyl)-1H-indazol-3-amine
Purity 95
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9673

Safety Information

signalword Danger
pcodes P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=9673
pictograms GHS06;GHS07
hstatements H301;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart