APO455834716 Apollo Scientific Ltd

3-Fluoro-[1,1'-biphenyl]-4-carbonitrile

  • MDL number MFCD06797374
  • Cas Number : 503177-15-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78-80
Synonym 4-Cyano-3-fluorobiphenyl, 2-Fluoro-4-phenylbenzonitrile
Purity 95
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10327

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=10327
pictograms GHS07;GHS09
hstatements H302;H312;H332;H319;H411
Availability details for
Quantity
  • Availability
  • Add to Cart