APO455834718 Apollo Scientific Ltd

4'-Fluoro-2-hydroxyacetophenone

  • MDL number MFCD02679035
  • Cas Number : 403-31-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113-117
Synonym 1-(4-Fluorophenyl)-2-hydroxyethan-1-one
Purity 95
Flash Point >110
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10328

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=10328
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart