APO455834720 Apollo Scientific Ltd

3,5-Bis(trifluoromethyl)-2,6-dichlorobenzal chloride

  • MDL number MFCD03094129
  • Cas Number : 501657-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1,5-Bis(trifluoromethyl)-2,4-dichloro-3-(dichloromethyl)benzene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5725

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=5725
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart