APO455834722 Apollo Scientific Ltd

3-Bromo-5-fluoroanisole

  • MDL number MFCD01861129
  • Cas Number : 29578-39-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Bromo-3-fluoro-5-methoxybenzene, 3-Bromo-5-fluorophenyl methyl ether
Purity 96
Density 1.59
Flash Point >90
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 94/17mm
Product Link https://store.apolloscientific.co.uk/product/3-bromo-5-fluoroanisole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-5-fluoroanisole
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart