APO455834726 Apollo Scientific Ltd

2-Chloro-3,4-difluoronitrobenzene

  • MDL number MFCD06658262
  • Cas Number : 169468-83-1
  • Molecular Weight : 193.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Chloro-3,4-difluoro-1-nitrobenzene
Purity 96
Flash Point 102
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-34-difluoronitrobenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-34-difluoronitrobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart