APO455834729 Apollo Scientific Ltd

5-Fluoro-2-methylpyridine

  • MDL number MFCD03095271
  • Cas Number : 31181-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Fluoro-2-picoline
Purity 98
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 114.7
Product Link https://store.apolloscientific.co.uk/product/5-fluoro-2-methylpyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-fluoro-2-methylpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart