APO455834739 Apollo Scientific Ltd

4,5-Dichloro-2-methylbenzotrifluoride

  • MDL number MFCD06246858
  • Cas Number : 74483-51-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4,5-Dichloro-2-(trifluoromethyl)toluene, 1,2-Dichloro-4-methyl-5-(trifluoromethyl)benzene
Flash Point 88
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 97-100/23mm
Product Link https://store.apolloscientific.co.uk/product/45-dichloro-2-methylbenzotrifluoride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/45-dichloro-2-methylbenzotrifluoride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart