APO455834740 Apollo Scientific Ltd

4-Bromophenyl trifluoromethyl sulphide

  • MDL number MFCD00040840
  • Cas Number : 333-47-1
  • Molecular Weight : 257.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 83
Iupac Name 1-bromo-4-[(trifluoromethyl)sulfanyl]benzene
density 1.71
Inchi Code 1S/C7H4BrF3S/c8-5-1-3-6(4-2-5)12-7(9,10)11/h1-4H
Inchi Key CQQBVMXLTBXNLB-UHFFFAOYSA-N

Description

Melting Point 83
Synonym 4-[(Trifluoromethyl)thio]bromobenzene, 1-Bromo-4-[(trifluoromethyl)thio]benzene
Purity 97%
Density 1.71
Flash Point 81/15mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromophenyl-trifluoromethyl-sulphide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromophenyl-trifluoromethyl-sulphide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart