APO455834743 Apollo Scientific Ltd

2-Chloro-3-fluorobenzeneboronic acid

  • MDL number MFCD07363741
  • Cas Number : 871329-52-1
  • Molecular Weight : 174.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 80-84
Iupac Name 2-chloro-3-fluorophenylboronic acid
Inchi Code 1S/C6H5BClFO2/c8-6-4(7(10)11)2-1-3-5(6)9/h1-3,10-11H
Inchi Key TYOGIUGNZOBBHE-UHFFFAOYSA-N

Description

Purity 99%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 80-84
Product Link https://store.apolloscientific.co.uk/product/2-chloro-3-fluorobenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-3-fluorobenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart