APO455834744 Apollo Scientific Ltd

3-Amino-4-(trifluoromethoxy)bromobenzene

  • MDL number MFCD04039242
  • Cas Number : 886762-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Bromo-2-(trifluoromethoxy)aniline
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-amino-4-trifluoromethoxybromobenzene

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-4-trifluoromethoxybromobenzene
pictograms GHS06
hstatements H301+H311+H331;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart