APO455834745 Apollo Scientific Ltd

5-Bromo-2-(trifluoromethoxy)nitrobenzene

  • MDL number MFCD04039243
  • Cas Number : 95668-20-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Bromo-2-nitro-1-(trifluoromethoxy)benzene, 4-Bromo-2-nitrophenyl trifluoromethyl ether, 4-Bromo-2-nitro-alpha,alpha,alpha-trifluoroanisole
Flash Point >110
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-trifluoromethoxynitrobenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-trifluoromethoxynitrobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart