APO455834746 Apollo Scientific Ltd

3-Nitro-4-(trifluoromethoxy)aniline

  • MDL number MFCD03840207
  • Cas Number : 2822-50-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90-92
Synonym 5-Amino-2-(trifluoromethoxy)nitrobenzene
Purity 98
Flash Point >110
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=6452

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=6452
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart