APO455834751 Apollo Scientific Ltd

2-Amino-5-bromo-2'-fluorobenzophenone

  • MDL number MFCD00038380
  • Cas Number : 1479-58-9
  • Molecular Weight : 294.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100-104
Synonym (2-Amino-5-bromophenyl)(2-fluorophenyl)methanone, 4-Bromo-2-(2-fluorobenzoyl)aniline
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-5-bromo-2-fluorobenzophenone

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-5-bromo-2-fluorobenzophenone
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart