APO455834752 Apollo Scientific Ltd

4-Bromo-alpha,alpha,beta,beta-tetrafluorophenetole

  • MDL number MFCD00042190
  • Cas Number : 68834-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Bromo-4-(1,1,2,2-tetrafluoroethoxy)benzene, 4-Bromophenyl 1,1,2,2-tetrafluoroethyl ether
Density 1.628
Flash Point 88
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 195-196
Product Link https://store.apolloscientific.co.uk/product/4-bromo-alphaalphabetabeta-tetrafluorophenetole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-alphaalphabetabeta-tetrafluorophenetole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart