APO455834753 Apollo Scientific Ltd

Ethyl perfluoro-2-bromopropanoate

  • MDL number MFCD00153721
  • Cas Number : 10186-73-9
  • Molecular Weight : 252.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Ethyl 2-bromo-2,3,3,3-tetrafluoropropionate, Ethyl 2-bromo-2,3,3,3-tetrafluoropropanoate
Purity 97
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 122-124
Product Link https://store.apolloscientific.co.uk/product/ethyl-perfluoro-2-bromopropanoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-perfluoro-2-bromopropanoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart