APO455834754 Apollo Scientific Ltd

2,3,4-Trifluorobromobenzene

  • MDL number MFCD00013248
  • Cas Number : 176317-02-5
  • Molecular Weight : 210.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Bromo-2,3,4-trifluorobenzene
Purity 99
Density 1.77
Flash Point 62
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 47-49/60mm
Product Link https://store.apolloscientific.co.uk/product/234-trifluorobromobenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/234-trifluorobromobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart