APO455834764 Apollo Scientific Ltd

2-Bromo-1,1,2-trifluoroethyl ethyl ether

  • MDL number MFCD00042465
  • Cas Number : 380-78-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Bromo-1-ethoxy-1,1,2-trifluoroethane, 1-Bromo-1,2,2-trifluoro-3-oxapentane
Purity 97
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 116
Product Link http://www.apolloscientific.co.uk/display_item.php?id=646

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=646
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart