APO455834768 Apollo Scientific Ltd

4-Bromo-1-iodo-2-(trifluoromethoxy)benzene

  • MDL number MFCD00179334
  • Cas Number : 175278-12-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 26
Synonym 5-Bromo-2-iodophenyl trifluoromethyl ether, 5-Bromo-2-iodo-alpha,alpha,alpha-trifluoroanisole
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 112-114/15mm
Product Link https://store.apolloscientific.co.uk/product/4-bromo-1-iodo-2-trifluoromethoxybenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-1-iodo-2-trifluoromethoxybenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart