APO455834769 Apollo Scientific Ltd

2'-Bromo-4'-(trifluoromethyl)acetanilide

  • MDL number MFCD00067782
  • Cas Number : 175135-49-6
  • Molecular Weight : 282.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144-146
Synonym 4-Acetamido-3-bromobenzotrifluoride, N-[2-Bromo-4-(trifluoromethyl)phenyl]acetamide
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-4-trifluoromethylacetanilide

Safety Information

signalword Warning
pcodes P280;P302+P352;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/2-bromo-4-trifluoromethylacetanilide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart