APO455834770 Apollo Scientific Ltd

4'-Bromo-3'-(trifluoromethyl)acetanilide

  • MDL number MFCD00052361
  • Cas Number : 41513-05-7
  • Molecular Weight : 282.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 116-119
Iupac Name N-[4-bromo-3-(trifluoromethyl)phenyl]acetamide
Inchi Code 1S/C9H7BrF3NO/c1-5(15)14-6-2-3-8(10)7(4-6)9(11,12)13/h2-4H,1H3,(H,14,15)
Inchi Key AOHSBSXIGFIMNC-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 116-119
Product Link https://store.apolloscientific.co.uk/product/4-bromo-3-trifluoromethylacetanilide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-3-trifluoromethylacetanilide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart