APO455834777 Apollo Scientific Ltd

1-Bromoundecafluorocyclohexane

  • MDL number MFCD00153732
  • Cas Number : 336-13-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Perfluoro-1-bromocyclohexane, Bromo(undecafluoro)cyclohexane
Purity 97
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 92/735mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=595

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=595
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart