APO455834781 Apollo Scientific Ltd

n-Butyl 1,1,2,2-tetrafluoroethyl ether

  • MDL number MFCD00042107
  • Cas Number : 358-37-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.1147
Flash Point 10
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 49/113mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=681

Safety Information

signalword Warning
pcodes P210;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=681
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart