APO455834784 Apollo Scientific Ltd

3-Chloro-5-fluorobenzeneboronic acid

  • MDL number MFCD06801741
  • Cas Number : 328956-61-2
  • Molecular Weight : 174.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 272-276
Iupac Name 3-chloro-5-fluorophenylboronic acid
Inchi Code 1S/C6H5BClFO2/c8-5-1-4(7(10)11)2-6(9)3-5/h1-3,10-11H
Inchi Key XYQDHVBKUNNLGZ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 272-276
Product Link https://store.apolloscientific.co.uk/product/3-chloro-5-fluorobenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-5-fluorobenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart