APO455834786 Apollo Scientific Ltd

2-Fluoro-5-(trifluoromethyl)benzeneboronic acid

  • MDL number MFCD03701538
  • Cas Number : 352535-96-7
  • Molecular Weight : 207.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104-109
Synonym 2-Fluoro-5-(trifluoromethyl)phenylboronic acid
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-5-trifluoromethylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-5-trifluoromethylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart