APO455834788 Apollo Scientific Ltd

3-Fluoro-4-hydroxybenzeneboronic acid

  • MDL number MFCD06659838
  • Cas Number : 182344-14-5
  • Molecular Weight : 155.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 252-256
Iupac Name 3-fluoro-4-hydroxyphenylboronic acid
Inchi Code 1S/C6H6BFO3/c8-5-3-4(7(10)11)1-2-6(5)9/h1-3,9-11H
Inchi Key OYNDLOJPYURCJG-UHFFFAOYSA-N

Description

Synonym 4-Borono-2-fluorophenol
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 252-256
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-4-hydroxybenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-4-hydroxybenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart