APO455834790 Apollo Scientific Ltd

2-Chloro-5-fluorobenzeneboronic acid

  • MDL number MFCD06656272
  • Cas Number : 444666-39-1
  • Molecular Weight : 174.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 78-82
Iupac Name 2-chloro-5-fluorophenylboronic acid
Inchi Code 1S/C6H5BClFO2/c8-6-2-1-4(9)3-5(6)7(10)11/h1-3,10-11H
Inchi Key OVTZJDSREVDCTJ-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 78-82
Product Link https://store.apolloscientific.co.uk/product/2-chloro-5-fluorobenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-5-fluorobenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart