APO455834794 Apollo Scientific Ltd

3-Cyano-4-fluorobenzeneboronic acid

  • MDL number MFCD03095130
  • Cas Number : 214210-21-6
  • Molecular Weight : 164.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >300
Synonym 5-Borono-2-fluorobenzonitrile
Purity 97
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-cyano-4-fluorobenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-cyano-4-fluorobenzeneboronic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart