APO455834800 Apollo Scientific Ltd

2-Fluoro-5-(methoxycarbonyl)benzeneboronic acid

  • MDL number MFCD06659868
  • Cas Number : 850568-04-6
  • Molecular Weight : 197.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210-212
Synonym Methyl 3-borono-4-fluorobenzoate, [(2-Fluoro-5-methoxycarbonyl)phenyl]boronic acid
Purity 95
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-5-methoxycarbonylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-5-methoxycarbonylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart