APO455834802 Apollo Scientific Ltd

2-Oxo-3'-(trifluoromethyl)propiophenone

  • MDL number MFCD01941304
  • Cas Number : 10557-15-0
  • Molecular Weight : 216.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 25-26
Synonym 1-[3-(Trifluoromethyl)phenyl]propane-1,2-dione
Purity 95
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 80/0.07mm
Product Link https://store.apolloscientific.co.uk/product/2-oxo-3-trifluoromethylpropiophenone

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-oxo-3-trifluoromethylpropiophenone
pictograms GHS07
hstatements H302;H315;H319;H332
Availability details for
Quantity
  • Availability
  • Add to Cart