APO455834803 Apollo Scientific Ltd

2-Fluoro-5-(trifluoromethyl)benzyl chloride

  • MDL number MFCD05864315
  • Cas Number : 883543-26-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(Chloromethyl)-1-fluoro-4-(trifluoromethyl)benzene, 3-(Chloromethyl)-4-fluorobenzotrifluoride
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9885

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=9885
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart