APO455834804 Apollo Scientific Ltd

3-Chloro-4-(trifluoromethyl)benzeneboronic acid

  • MDL number MFCD03788417
  • Cas Number : 847756-88-1
  • Molecular Weight : 224.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206-212
Synonym 4-Borono-2-chlorobenzotrifluoride
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloro-4-trifluoromethylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-4-trifluoromethylbenzeneboronic-acid
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart