APO455834808 Apollo Scientific Ltd

3-Bromo-2-fluorophenol

  • MDL number MFCD09864673
  • Cas Number : 156682-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 52-54
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=40857

Safety Information

signalword Warning
pcodes P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=40857
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart