APO455834811 Apollo Scientific Ltd

N-(Chloroacetyl)-3-fluoroaniline

  • MDL number MFCD01034343
  • Cas Number : 350-81-2
  • Molecular Weight : 187.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121-123
Synonym 2-Chloro-3'-fluoroacetanilide
Purity 95
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/n-chloroacetyl-3-fluoroaniline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/n-chloroacetyl-3-fluoroaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart