APO455834812 Apollo Scientific Ltd

N-(Chloroacetyl)-4-fluoroaniline

  • MDL number MFCD00015730
  • Cas Number : 351-04-2
  • Molecular Weight : 187.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131-132
Synonym 2-Chloro-4'-fluoroacetanilide, 2-Chloro-N-(4-fluorophenyl)acetamide
Purity 97
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/n-chloroacetyl-4-fluoroaniline

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/n-chloroacetyl-4-fluoroaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart