APO455834815 Apollo Scientific Ltd

2,6-Difluoro-3-nitrobenzoic acid

  • MDL number MFCD00119291
  • Cas Number : 83141-10-0
  • Molecular Weight : 203.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 92-96
Iupac Name 2,6-difluoro-3-nitrobenzoic acid
Inchi Code 1S/C7H3F2NO4/c8-3-1-2-4(10(13)14)6(9)5(3)7(11)12/h1-2H,(H,11,12)
Inchi Key PDDHSNODMFIIRV-UHFFFAOYSA-N

Description

Synonym 3-Carboxy-2,4-difluoronitrobenzene
Purity 98%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 92-96
Product Link https://store.apolloscientific.co.uk/product/26-difluoro-3-nitrobenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/26-difluoro-3-nitrobenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart