APO455834816 Apollo Scientific Ltd

4-Chloro-2-fluoro-5-nitrobenzoic acid

  • MDL number MFCD04115725
  • Cas Number : 35112-05-1
  • Molecular Weight : 219.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146-153
Synonym 5-Carboxy-2-chloro-4-fluoronitrobenzene
Purity 98
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-chloro-2-fluoro-5-nitrobenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloro-2-fluoro-5-nitrobenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart