APO455834841 Apollo Scientific Ltd

2-Fluorophenylhydrazine

  • MDL number MFCD00041260
  • Cas Number : 2368-80-1
  • Molecular Weight : 126.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 47
Iupac Name 1-(2-fluorophenyl)hydrazine
Inchi Code 1S/C6H7FN2/c7-5-3-1-2-4-6(5)9-8/h1-4,9H,8H2
Inchi Key PENWGQNPFRRVQI-UHFFFAOYSA-N

Description

Flash Point 76-79/3mm
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 47
Product Link https://store.apolloscientific.co.uk/product/2-fluorophenylhydrazine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluorophenylhydrazine
pictograms GHS07
hstatements H302+H312+H332;H319
Availability details for
Quantity
  • Availability
  • Add to Cart