APO455834844 Apollo Scientific Ltd

4-Fluoro-3-(trifluoromethyl)benzeneboronic acid

  • MDL number MFCD07363787
  • Cas Number : 182344-23-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-174
Synonym 5-Borono-2-fluorobenzotrifluoride
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-3-trifluoromethylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-3-trifluoromethylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart