APO455834847 Apollo Scientific Ltd

Methyl 2-bromo-5-fluorobenzoate

  • MDL number MFCD00086167
  • Cas Number : 6942-39-8
  • Molecular Weight : 233.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-2-bromo-5-fluorobenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2-bromo-5-fluorobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart