APO455834854 Apollo Scientific Ltd

2,6-Dichloro-3-fluorobenzoic acid

  • MDL number MFCD06658165
  • Cas Number : 178813-78-0
  • Molecular Weight : 209

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/26-dichloro-3-fluorobenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/26-dichloro-3-fluorobenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart