APO455834858 Apollo Scientific Ltd

2-Chloroethyl 3-[(trifluoromethyl)phenyl] sulphone

  • MDL number MFCD00068016
  • Cas Number : 175203-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 52-55
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=770

Safety Information

signalword Danger
pcodes P280;P305+P351+P338
MSDS http://www.apolloscientific.co.uk/display_item.php?id=770
pictograms GHS05;GHS07
hstatements H302;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart