APO455834862 Apollo Scientific Ltd

6-(Trifluoromethyl)nicotinamide

  • MDL number MFCD01862646
  • Cas Number : 386715-35-1
  • Molecular Weight : 190.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185-187
Synonym 6-(Trifluoromethyl)pyridine-3-carboxamide, 5-Carbamoyl-2-(trifluoromethyl)pyridine
Purity 99
Flash Point >110
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-trifluoromethylnicotinamide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-trifluoromethylnicotinamide
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart