APO455834863 Apollo Scientific Ltd

3'-Chloro-4'-fluoroacetophenone

  • MDL number MFCD00042203
  • Cas Number : 2923-66-2
  • Molecular Weight : 172.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 40-42
Synonym 1-(3-Chloro-4-fluorophenyl)ethan-1-one
Purity 99
Physical Form Solid-Fused
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 126-128/15mm
Product Link https://store.apolloscientific.co.uk/product/3-chloro-4-fluoroacetophenone

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-4-fluoroacetophenone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart