APO455834901 Apollo Scientific Ltd

2-Chloro-6-fluorobenzoyl chloride

  • MDL number MFCD00178746
  • Cas Number : 79455-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.43
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 106-108/15mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=822

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=822
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart