APO455834903 Apollo Scientific Ltd

2-Chloro-4-fluorobenzylamine

  • MDL number MFCD00042532
  • Cas Number : 15205-11-5
  • Molecular Weight : 159.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (2-Chloro-4-fluorophenyl)methylamine
Purity 97
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 94-96/15mm
Product Link https://store.apolloscientific.co.uk/product/2-chloro-4-fluorobenzylamine

Safety Information

signalword Danger
pcodes P260;P280;P310
MSDS https://store.apolloscientific.co.uk/product/2-chloro-4-fluorobenzylamine
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart